Tisk…
Scoops, long handled, SteriWare®
Dodavatel: Sampling Systems
8341H-100S 8341H-100 8341H-25 8341H-25S 8341H-50 8341H-50S 8341R-100S
SAMP8341H-100SEA 8400 CZK
SAMP8341H-100S SAMP8341H-100 300-0580 300-0581 300-0582 300-0583 300-0584
Scoops, long handled, SteriWare®
Lopatky
These SteriWare® long handled scoops are the ideal tool for reaching into deep containers. The extra long handle will help operators keep their hands out of the product.
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • Available sterile (SAL 10-6 ) and non sterile
They are all manufactured and packed in a cleanroom to ensure cleanliness and reduce particulate contamination.
These single use scoops are ideal for use in the laboratory, production and warehouse.
Handle length: 300 mm
Certifikace: Made fromMore Product Information