Tisk…
Sample Lance
Dodavatel: Sampling Systems
1660H-1000 1660H-600 1660P-1000 1660P-600 1665H-1000 1665H-600 1665P-1000 1665P-600
300-0448EA 20500 CZK
300-0448 300-0449 300-0450 300-0451 300-0452 300-0453 300-0454 300-0455
Sample Lance
Vzorkovače Vzorkovače pevné
The Sample Lance is excellent for taking cross sectional samples of powders and some pastes.
  • Easy to use
  • Great for hard and dense powders
  • Available with a range of different handles
  • Made from high quality 316L stainless steel
The Sample Lance is excellent for sampling powders and pastes. Various models allow for good cross sectional sampling of powders with varying properties. If the powder is hard and dense, then the addition of the heavy-duty handle is a convenient feature.