Tisk…
Detectable volumetric spoon, blue
Dodavatel: BURKLE
5378-4087 5378-4099 5378-4085 5378-4091 5378-4093 5378-4097 5378-4089 5378-4095
300-0397EA 1810 CZK
300-0397 300-0405 300-0396 300-0399 300-0402 300-0404 300-0398 300-0403
Detectable volumetric spoon, blue
Lžíce
These samplers are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.
  • PS detectable, blue
  • Individually packaged and sterilised
  • Can be detected by colour scanners, metal detectors or X-rays
  • Can be used as part of HACCP/IFS foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • With stripper edge
  • For single use
More Product Information