Tisk…
FEP Fluoropolymer Gas and Liquid Sampling Bags
Dodavatel: Saint Gobain Life Sciences
D1075005 D1075001 D1075004 D1075006
300-0275EA 2660 CZK
300-0275 300-0271 300-0274 300-0276
FEP Fluoropolymer Gas and Liquid Sampling Bags
Sáčky Sáčky na vzorkování plynů
These flexible, sturdy bags, made from 5 mil thick FEP fluoropolymer film, are heatsealed on all four sides for complete chemical resistance and leakproof operation. Can be used for gas sample analysis, extracting air samples from smokestack exhaust systems, monitoring air quality or calibrating instruments in the field.
  • Easy to use
  • Flexible and sturdy
  • Completely chemical-resistant
  • Wil not contaminate samples
  • Leak proof, heat sealed all four sides
  • Completely chemical-resistant
  • Low permeability – no sample loss
  • No aging – unlimited storage life
  • Unique seam
More Product Information