Tisk…
Spoons, PharmaSpoon
Dodavatel: BURKLE
5324-0001
300-0075EA 17600 CZK
300-0075
Spoons, PharmaSpoon
Lžíce
Stainless steel V4A (1.4404), high polish finish. For taking samples of powdery substances, intermediate products and material in the pharmaceutical industry.
  • Possible residues are even visible in the trace sector (µg sector)
  • Also suitable for sterile sampling
  • Made of materials with excellent surface qualities to prevent cross-contamination