Tisk…
Conical hand drill, QualiRod
Dodavatel: BURKLE
5311-0001 5311-0002
300-0018EA 2220 CZK
300-0018 300-0140
Conical hand drill, QualiRod
Vzorkovače Vzorkovače pevné
Conical hand drill with sharp cutting edges and a solid handle made from stainless steel V2A (1.4301)/AISI 304.
  • Sterilisable
Designed to cut out samples from soft, paste-like substances (cheese, butter, clay, loam, soap and wax).
Certifikace: ISO 707