Tisk…

o-Xylén 98%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™

Dodavatel: Honeywell Chemicals
Nebezpečí

295884-1L
295884-1LEA 3720 CZK
295884-1L
o-Xylén 98%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
o-Xylén

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Vzorec: C₈H₁₀
Bod varu: 143-145 °C (1,013 hPa)
Bod tání: -25 °C
Hustota: 0,88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008519
CAS číslo: 95-47-6
EINECS: 202-422-2
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dd6c62d1c62b6e7f71df7e157aae77162b3bcf1&vid=3ec8c09bb94f634e9f90b6f9e8320cd626f276d0&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay (GC) Min. 97.5 %
Non-volatile matter Max. 0.0003 %
Water (Karl Fischer) Max. 0.02 %
Identity (IR) complying
Absorbance at 350 nm Max. 0.01
Absorbance at 325 nm Max. 0.05
Absorbance at 300 nm Max. 0.40
Absorbance at 288 nm Max. 1.00
Autoignition Temperature 867 °F
Vapor Pressure <0.1 atm ( 21.1 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR