Tisk…
Dual Lock™ SJ354D Reclosable Fastener Twin Pack
Dodavatel: 3M
SJ354B SJ354D
293-0005EA 1460 CZK
293-0005 293-0014
Dual Lock™ SJ354D Reclosable Fastener Twin Pack
Pojidla
This reclosable fastener has a rubber based adhesive on the back of a black, polypropylene reclosable fastener. It's suitable for bonding many plastics, powder coated paints, and other low surface energy materials.
  • Snap-in-place installation replaces screws, nails, snaps, and rivets
  • Type 250 mates to Type 170, 250, or 400
  • Closes with an audible snap
  • Opens and closes 1000 times before losing 50% of original tensile strength
  • Pressure-sensitive adhesive provides excellent initial tack to most surfaces
  • Helps reduce vibration
Designed for use with manyMore Product Information