Tisk…
Dispensers for Parafilm® M sealing film, Nalgene®
Dodavatel: Thermo Fisher Scientific
5833-0001
291-0013EA 4630 CZK
291-0013
Dispensers for Parafilm® M sealing film, Nalgene®
Zásobníky na fólie/parafilm
For clean storage and easy dispensing of Parafilm® M sealing film.
  • Designed for two rolls 50 mm wide or one roll 100 mm wide
  • Easy to clean
  • Clear acrylic