Tisk…
Hydrophobic interaction chromatography columns, HiTrap™ Capto™ HIC selection kit
Dodavatel: Cytiva
29-3210-87
29-3210-87EA 22460 CZK
29-3210-87
Hydrophobic interaction chromatography columns, HiTrap™ Capto™ HIC selection kit
Kolony chromatografické
The Capto hydrophobic interaction chromatography (HIC) selection kit has a set of five 1 ml ready-to-use HiTrap™ columns for resin screening and small-scale protein purification. The HiTrap™ Capto HIC selection kit is useful in process development or research applications.