Tisk…
Affinity chromatography columns, His SpinTrap™ TALON®
Dodavatel: Cytiva
29-0005-93
29-0005-93EA 13120 CZK
29-0005-93
Affinity chromatography columns, His SpinTrap™ TALON®
Kolony chromatografické
His SpinTrap™ TALON® is a single-use column designed for high purity, small-scale purification and screening of histidine-tagged proteins from clarified cell lysates.
  • Suitable for purification of multiple samples in parallel, such as screening
  • With a microcentrifuge, one run takes about 10 minutes
Convenient, ready-to-use single-use spin column, prepacked with 100 μl TALON® Superflow™ cobalt-IMAC (immobilized metal affinity chromatography) medium offering a different selectivity compared to nickel-charged media.
The columns make parallelMore Product Information
Objednat
Produktová specifikace
Environmentally Preferable
Kapacita vázání up to 1 mg histidine-tagged