Tisk…

Anion exchange chromatography column, HiScreen™ Capto™ Q

Dodavatel: Cytiva
28-9269-78
28-9269-78EA 5030 CZK
28-9269-78
Anion exchange chromatography column, HiScreen™ Capto™ Q
Kolony chromatografické
HiScreen Capto Q is a column packed with a strong anion exchange modern resin, combining speed and capacity. Suitable for process development.

  • Prepacked with Capto Q strong anion exchange chromatography resin
  • Convenient method optimization and parameter screening because of the 10 cm bed height
  • These columns are easily connected in series to increase bed height to 20 cm
  • Fast results and minimal consumption of sample and buffer consumption through the small bed volume
  • BioProcess columns packed with the same chromatography resin and using the same linear
More Product Information
Objednat

Produktová specifikace

Environmentally Preferable
Maximální tlak 3 bar
Rozsah pH 2 - 12
Uskladnění 4 to 30 °C, 20% Ethanol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR