Tisk…
dNTP solutions, illustra™
Dodavatel: Cytiva
28-4065-42 28-4065-41 28-4065-32 28-4065-22 28-4065-02 28-4065-01 28-4065-12
28-4065-42EA 5000 CZK
28-4065-42 28-4065-41 28-4065-32 28-4065-22 28-4065-02 28-4065-01 28-4065-12
dNTP solutions, illustra™
Činidla pro nukleové kyseliny Nukleotidy
illustra™ dNTPs are high purity solutions for enzymatically sensitive applications, such as nucleic acid amplification, sequencing, labelling, mutagenesis, cDNA synthesis and expression profiling.
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
Objednat
Produktová specifikace
Environmentally Preferable
Aplikace