Tisk…
Multimode columns, HiTrap™ Capto™ adhere
Dodavatel: Cytiva
28-4058-46 28-4058-44 29-4004-56
28-4058-46EA 30060 CZK
28-4058-46 28-4058-44 29-4004-56
Multimode columns, HiTrap™ Capto™ adhere
Kolony chromatografické
HiTrap Capto adhere is prepacked with Capto adhere, a multimodal BioProcess ion exchange resin designed to remove contaminats in post-protein A purification.
  • High capacity and productivity in flowthrough mode
  • Savings in time and operating costs with a two-step chromatographic process
  • Wide operational window of pH and conductivity
  • Prepacked HiTrap columns for easy screening and convenient process development
Capto adhere is a strong anion exchanger with multimodal functionality. The multimodal functionality gives a different selectivity compared toMore Product Information