Tisk…
Affinity chromatography columns, His SpinTrap™
Dodavatel: Cytiva
28-4013-53
28-4013-53EA 13880 CZK
28-4013-53
Affinity chromatography columns, His SpinTrap™
Kolony chromatografické
His SpinTrap is a prepacked, single-use spin column for purifying histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).
  • For simple, small-scale purification of histidine-tagged* proteins and rapid expression screening
  • Ni Sepharose High Performance in prepacked spin columns
  • Yields up to 750 µg pure protein per column
  • Allows direct purification of both clarified and unclarified cell lysates
Purification of unclarified samples on His SpinTrap columns minimizes loss of target protein caused by manual operations such as sampleMore Product Information