Tisk…

Chlorid sodný 99.5-100.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent

Dodavatel: VWR Chemicals

27810.295EA 1210 CZK
27810.295 27810.364 27810.262 27810.460
Chlorid sodný 99.5-100.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Chlorid sodný
Vzorec: NaCl
Bod varu: 1413 °C (1013 hPa)
Bod tání: 801 °C
Hustota: 2,16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS číslo: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Objednat

Výsledky specifikačního testu

Assay 99.5 - 100.5 %
Appearance of Solution S Passes test Ph.Eur.
Identification A Passes test Ph.Eur.
Identification B Passes test Ph.Eur.
NO2 (Nitrite) Passes test Ph.Eur.
Solution in water Passes test
Acidity or alkalinity Max. 0.0005 meq/g
pH (25°C; 5 %) 5.0 - 9.0
Heavy metals (as Pb) Max. 2 ppm
Hexacyanoferrates (as Fe(CN)6) Max. 1 ppm
Insolubility in water Max. 50 ppm
Loss on drying (105°C) Max. 0.5 %
Magnesium and alkaline earth metals Max. 100 ppm
Total N (Nitrogen) Max. 5 ppm
Br (Bromide) Max. 50 ppm
ClO3 + NO3 (as NO3) Max. 30 ppm
I (Iodide) Max. 10 ppm
PO4 (Phosphate) Max. 5 ppm
SO4 (Sulphate) Max. 40 ppm
As (Arsenic) Max. 0.4 ppm
Ba (Barium) Max. 10 ppm
Ca (Calcium) Max. 20 ppm
Cu (Copper) Max. 2 ppm
Fe (Iron) Max. 1 ppm
K (Potassium) Max. 50 ppm
Mg (Magnesium) Max. 10 ppm
Ni (Nickel) Max. 10 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR