Tisk…

Octan sodný ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent

Dodavatel: VWR Chemicals

Synonyms: Acetic acid sodium salt

27653.235EA 1020 CZK
27653.235 27653.292 27653.260 27653.460 27653.360
Octan sodný ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Octan sodný
Vzorec: H₃CCOONa
MW: 82.03
Bod varu: 881.4 °C (1013 hPa)
Bod tání: 324
Hustota: 1.528
Bod vzplanutí: >250 °C (ot. pohárek)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS číslo: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Objednat

Výsledky specifikačního testu

Assay Min. 99.0 %
Identification Passes test
IR Spectrum Passes test
pH (25°C; 5 %) 7.0 - 9.2
Heavy metals (as Pb) Max. 10 ppm
Insolubility in water Max. 100 ppm
Loss on drying (120°C) Max. 1.0 %
Cl (Chloride) Max. 20 ppm
PO4 (Phosphate) Max. 10 ppm
SO4 (Sulphate) Max. 30 ppm
Al (Aluminium) Max. 10 ppm
Ca (Calcium) Max. 50 ppm
Cu (Copper) Max. 3 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 0.05 %
Mg (Magnesium) Max. 20 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR