Tisk…
Flasks, Erlenmeyer, with standard ground joint
Dodavatel: Lenz Laborglas GmbH & CO.KG
03021413 03021422 03021428 03021437 03021922 03021928 03021937 03021946 03021949 03021952 03022428 03022437 03022446 03022449 03022452 03022458 03022470 03022473 03022928 03022937 03022946 03022949 03022952 03022958 03022970 03022973 03024546 03024549 03024552 03024558 03024570 03024573 03024582 03024576 03022943 03021943
201-0180EA 429 CZK
201-0180 201-1101 201-1102 201-1103 201-0315 201-1121 201-1123 201-0183 201-0316 201-0185 201-0317 201-1119 201-1122 201-1120 201-0318 201-0319 201-0186 201-0187 201-1112 201-1113 201-1114 201-1115 201-1116 201-1117 201-1118 271-1119 201-0636 271-1125 271-1126 271-1127 271-1128 271-1129 201-0638 201-0637 201-0635 LENZ03021943
Flasks, Erlenmeyer, with standard ground joint
Baňky Baňky dle Erlenmeyera
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area.
DIN 12387 (except Cat. No. 201-1114, 201-1122, 201-0318, 201-1116 and 271-1126)
Certifikace: DIN 12387 (except Cat. No. 201-1114, 201-1122, 201-0318, 201-1116 and 271-1126)