Tisk…
Adapters, receiver, with side socket, Quickfit®
Dodavatel: SciLabware
RA4/11
271-0012EA 1870 CZK
271-0012
Adapters, receiver, with side socket, Quickfit®
Komponenty pro destilaci Adaptéry destilační
PYREX®, Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With side socket inclined at an angle of 105°.