Tisk…
Deaza dGTP
Dodavatel: Cytiva
27-2090-02
27-2090-02EA 10330 CZK
27-2090-02
Deaza dGTP
Činidla pro nukleové kyseliny Nukleotidy
Band compressions are ambiguous band patterns on sequencing gels caused by stable intra-strand secondary structures that are not fully denatured during electrophoresis. Their occurrence can be reduced by substituting nucleotide analogues in the sequencing mixes. 7-Deaza dGTP (c⁷dGTP) can be used to resolve G-band compressions.
  • For reducing “band compressions” in sequencing gels
  • Supplied as a 5 mM solution