Tisk…

Manganistan draselný 99.0-100.5%, krystalický Ph. Eur.

Dodavatel: VWR Chemicals
Nebezpečí

26904.293EA 2510 CZK
26904.293 26904.464
Manganistan draselný 99.0-100.5%, krystalický Ph. Eur.
Manganistan draselný
Vzorec: KMnO₄
Bod tání: > 240 °C
Hustota: 2,7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS číslo: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II
Merck Index: 12,07824

Objednat

Výsledky specifikačního testu

Assay 99.0 - 100.5 %
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification (K) Passes test
Appearance of solution Passes test
Substances insoluble in water ≤ 1.0 %
Cl (Chloride) ≤ 200 ppm
SO₄ (Sulphate) ≤ 500 ppm
Water ≤ 0.5 %
Residual solvents Passes test
As (Arsenic) ≤ 1 ppm
Cd (Cadmium) ≤ 1 ppm
Co (Cobalt) ≤ 1 ppm
Hg (Mercury) ≤ 1 ppm
Ni (Nickel) ≤ 1 ppm
Pb (Lead) ≤ 1 ppm
V (Vanadium) ≤ 1 ppm
Conforms to Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR