Tisk…

Uhličitan draselný 99,0, AnalaR NORMAPUR® zA Na_free

Dodavatel: VWR Chemicals
Varování

Synonyms: Potash

26727.267EA 1440 CZK
26727.267
Uhličitan draselný 99,0, AnalaR NORMAPUR® zA Na_free
Uhličitan draselný
Vzorec: K₂CO₃
MW: 138.21
Bod tání: 891
Hustota: 2.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS číslo: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay (calculated on dried substance) Min. 99.0 %
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 50 ppm
Loss on drying (300 °C) Max. 1 %
Substances precipitated by NH4OH Max. 100 ppm
Total N (Nitrogen) Max. 10 ppm
Total S (as SO4) Max. 30 ppm
Cl (Chloride) Max. 30 ppm
PO4 (Phosphate) Max. 10 ppm
SiO4 (as SiO2) Max. 50 ppm
Ca (Calcium) Max. 20 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 5 ppm
Mg (Magnesium) Max. 10 ppm
Na (Sodium) Max. 0.02 %
Pb (Lead) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR