Tisk…

parafín, Solidification point: 58-60°C, granules, TECHNICAL

Dodavatel: VWR Chemicals

Synonyms: Wax Paraffin, Paraffin wax

26154.291EA 3360 CZK
26154.291 26154.460
parafín, Solidification point: 58-60°C, granules, TECHNICAL
Parafín
Bod tání: 58…60 °C
Hustota: 0.9 g/cm³ (20 °C)
CAS číslo: 8002-74-2
EINECS: 232-315-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR