Tisk…

Murexide zA

Dodavatel: VWR Chemicals

Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt

25717.120EA 3550 CZK
25717.120
Murexide zA
Murexide
Vzorec: C₈H₈N₆O₆
MW: 284.19
Bod tání: 300
Storage Temperature: Ambient
MDL Number: MFCD00012777
CAS číslo: 3051-09-0
EINECS: 221-266-6
Merck Index: 13,06330

Objednat

Výsledky specifikačního testu

Suited for complexometry indicator Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR