Tisk…
Sequencing reaction purification, illustra™ AutoScreen 96 Well Plates
Dodavatel: Cytiva
25-9005-98
25-9005-98EA 31650 CZK
25-9005-98
Sequencing reaction purification, illustra™ AutoScreen 96 Well Plates
Činidla pro nukleové kyseliny Činidla Sekvenační
AutoScreen 96 well plate consists of a 96-well filter plate containing DNA grade Sephadex™ G-50 for purification of sequencing reactions prior to analysis on MegaBACE 1000 and can be used for other size exclusion applications.