Tisk…

n-Hexan 95, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA

Dodavatel: VWR Chemicals
Nebezpečí

Synonyms: Hexyl hydride, Hexanen, Hexane

24577.298EA 2900 CZK
24577.298 24577.323 24577.367 24577.460
n-Hexan 95, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA
n-Hexan
Vzorec: H₃C(CH₂)₄CH₃
MW: 86.18
Bod varu: 69
Bod tání: –94,3 °C
Hustota: 0.659 g/cm³ (20 °C)
Bod vzplanutí: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS číslo: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
REACH: 01-2119480412-44
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay (n-Hexane) Min. 95.0 %
Assay (Isomers) Min. 98.5 %
IR Spectrum Passes test
Thiophene Passes test
Acidity Max. 0.0002 meq/g
Boiling point 68 - 69 °C
Bromine value Max. 0.5
Colouration Max. 10 APHA
Density (20/4) 0.658 - 0.662
Density (20/20) 0.659 - 0.663
Refractive index (20°C) 1.375 - 1.376
Substances coloured by H2SO4 Max. 90 APHA
Benzene Max. 100 ppm
Evaporation residue Max. 10 ppm
Total S (as SO4) Max. 10 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.1 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.01 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR