Tisk…
VWR® Talon®, Clamps, Three Pronged
Dodavatel: VWR Collection
241-0404EA 3210 CZK
241-0404 241-0405 241-0406
VWR® Talon®, Clamps, Three Pronged
Svorky Svorky pro běžné účely
Stainless steel or nickel-plated zinc. These clamps securely hold every type of glassware and apparatus.
  • Extension arms allow placement of apparatus at various distances from lab frames without compromising the integrity of the experiment
  • Stainless steel clamps: Exceptional chemical resistance, clamps are electropolished and autoclavable
  • Nickel-plated zinc clamps: High tensile strength
  • Supplied with non-slip vinyl sleeves, and fiberglass covers for temperatures above 100 °C
Long, seamless tubing attaches clamp head securely and offers easy positioningMore Product Information