Tisk…

Silikagel, Davisil® Grade 62 pore size 150 Å, 35-60 mesh, high purity, Supelco®

Dodavatel: Merck

236845-100G 236845-1KG
236845-100GEA 3020 CZK
236845-100G 236845-1KG
Silikagel, Davisil® Grade 62 pore size 150 Å, 35-60 mesh, high purity, Supelco®
Silikagel
Vzorec: SiO₂
Bod varu: 2230 °C
Bod tání: > 1600 °C
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS číslo: 63231-67-4
EINECS: 231-545-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc93c274583970dc8baa52abfe2cf9ddb1f7c5d9&vid=9bbeb82d81a0a3c04adfd63ae4db4fd0eb202793&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR