Tisk…
Scalpels
Dodavatel: HAMMACHER KARL
HWB220-30 HWB221-35 HWB222-40 HWB224-35 HWB225-40 HWB223-30
233-0092EA 2380 CZK
233-0092 233-0093 233-0094 233-0095 HAMMHWB225-40 HAMMHWB223-30
Scalpels
Skalpely a skalpelové sady Skalpely
These scalpels are made from Wironit steel (18/12 stainless steel with 18% chrome and from 10 to 12% nickel).
  • Resistant to corrosion and anti-magnetic
  • With fixed blade and metal handle