Tisk…
Measuring scoops, Semadeni®
Dodavatel: SEMADENI
0042 0041 0044 0044_STERIL
231-0121EA 362 CZK
231-0121 SEMA0041 SEMA0044 SEMA0044_STERIL
Measuring scoops, Semadeni®
Lopatky
Measuring scoops made from white HDPE.
Solid, unbreakable and chemical resistant
With flat, bevelled shovel end
Suitable for food contact