Tisk…
Flow cytometry fixation/permeabilisation kit
Dodavatel: Biotium
23006
23006.EA 1420 CZK
23006.
Flow cytometry fixation/permeabilisation kit
Test Buněčná stanovení
Flow Cytometry Fixation/Permeabilisation Kit contains optimally formulated buffers for fixation and permeabilisation of suspension cells for immunofluorescence staining of intracellular antigens for analysis by flow cytometry. The bufffers are also available separately.
Informace o dodávce: Kit contains Fixation Buffer, 5 ml and Permeabilisation Buffer, 5 ml.