Tisk…

Triizopropanolát hlinitý ≥99.99% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Aluminiumtriisopropylate, Aluminiumisopropylate, Aluminiumtriisopropoxide, Aluminium isopropoxide

22982.14 22982.22
22982.14EA 4260 CZK
22982.14 22982.22
Triizopropanolát hlinitý ≥99.99% (metals basis)
Triizopropanolát hlinitý

Purity: 99.99+% CAS No.: 555-31-7 Molecular Formula: Al[OCH(CH3)2]3 Formula Weight: 204.25 Physical Form: Powder Density: 1.035 Refractive Index: 1.0346 Sensitivity: Moisture Sensitive Melting Point: 131-135° Boiling Point: 135°/10mm Merck Reference: 14,347 UN No.: 3181 MDL No.: MFCD00008870

Vzorec: C₉H₂₁AlO₃
MW: 204.25
Bod varu: 135 °C (10 mmHg)
Bod tání: 131…135 °C
Hustota: 1.035
Storage Temperature: Ambient
MDL Number: MFCD00008870
CAS číslo: 555-31-7
EINECS: 209-090-8
UN: 3181
ADR: 4.1 II
Merck Index: 14,347

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR