Tisk…

Síra 1000 mg/L ve vodě (from (NH₄)₂SO₄) Specpure® standard pro AAS

Dodavatel: Thermo Fisher Scientific

22958.AE 22958.AP
22958.AEEA 1450 CZK
22958.AE 22958.AP
Síra 1000 mg/L ve vodě (from (NH₄)₂SO₄) Specpure® standard pro AAS
Standardy Standardy kationické Standardy - síra

Molecular Formula: (NH₄)₂SO₄ in H₂O
Physical Form: Liquid
MDL No.: MFCD00085316


Certifikace: Traceable to NIST

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR