Tisk…
Pump tubing, Tygon® Chemical
Dodavatel: Saint Gobain Life Sciences
AD300007 AD300012 AD300017 AD300027 AD300038
228-0691EA 13550 CZK
228-0691 228-0692 228-0693 228-0694 228-0695
Pump tubing, Tygon® Chemical
Hadičky Přenosová hadička
Tygon® tubing, cream coloured.
  • Excellent resistance to acids, bases, salts, ketones and alcohols
  • No absorption of liquids
  • Extremely flexible and durable
Tygon® Chemical tubing is a co-extrusion developed to provide a combination of chemical resilience and durability in peristaltic pumps. The ultra-smooth inner core contains no plasticisers and resists absorption/adsorption of aqueous liquids. Ideal for applications involving sensitive media. The tubing can be autoclaved up to five times without impairing its service life.
Temperature resistance: –60More Product Information