Tisk…

Chloroform, Wasser_0,005 99,5 Stabilised, AnalaR NORMAPUR® zA

Dodavatel: VWR Chemicals
Nebezpečí

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: 2-Methyl-2-buten

22709.292EA 2950 CZK
22709.292
Chloroform, Wasser_0,005 99,5 Stabilised, AnalaR NORMAPUR® zA
Chloroform
Vzorec: CHCl₃
MW: 119.38
Bod varu: 61.7 °C (1013 hPa)
Bod tání: –63 °C
Hustota: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS číslo: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 01-2119486657-20
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay (calculated on anhydrous) Min. 99.5 %
Colour value Max. 10 APHA
Acidity Max. 0.0005 %
Residue on evaporation Max. 0.0005 %
Water (K.F.) Max. 0.0050 %
Dichloromethane Max. 0.0100 %
Carbon tetrachloride Max. 0.0100 %
Trichloroethene Max. 0.0100 %
1,1,2,2-Tetrachlorethane Max. 0.0100 %
Formaldehyde-sulphuric colouration Max. 60 APHA
Substances discoloured by H2SO4 Max. 35 APHA
Free chlorine Max. 0.00003 %
Cl (Chloride) Max. 0.5 ppm
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.05 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR