Tisk…

Uhličitan vápenatý ≥99%, GPR RECTAPUR®, precipitated

Dodavatel: VWR Chemicals

Synonyms: Marble

22296.294EA 2690 CZK
22296.294
Uhličitan vápenatý ≥99%, GPR RECTAPUR®, precipitated
Uhličitan vápenatý

  • White odorless powder
  • Nearly insoluble in water and alcohol
  • Soluble in acetic acid, hydrochloric acid and nitric acid
  • Median particle size d50 1.0 µm

Vzorec: CaCO₃
MW: 100.09
Bod tání: 825
Hustota: 2.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS číslo: 471-34-1
EINECS: 207-439-9
REACH: 01-2119486795-18
Merck Index: 13,01658

Objednat

Výsledky specifikačního testu

Assay Min. 99 %
Heavy metals (as Pb) Max. 20 ppm
Cl (Chloride) Max. 0.03 %
SO₄ (Sulphate) Max. 0.05 %
Fe (Iron) Max. 0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR