Tisk…
Funnel for conical ground joints
Dodavatel: VITLAB
70594 70694 70494
221-1409EA 1680 CZK
221-1409 221-1410 221-1418
Funnel for conical ground joints
Nálevky Nálevky filtrační Nálevky pro kónické závity
PP, translucent.
  • Ideal for transferring liquid or powdered reagents to multiple-neck flasks
  • Very good chemical resistance
  • Autoclavable
The funnel body is flattened on the side and the end of the shaft is shaped to fit conical ground joints.