Tisk…
Stoppers, with drip tip, MBL®
Dodavatel: SciLabware
TRF691 TRF692 TRF693 TRF695 TRF696 TRF697 TRF690 TRF698
217-0745EA 1510 CZK
217-0745 BIBBTRF692 217-0746 217-0747 217-0748 217-0749 BIBBTRF690 BIBBTRF698
Stoppers, with drip tip, MBL®
Zátky Zátky skleněné
Ideally suited for use with volumetric flasks etc. Lower closed end of stopper features a 'drip-tip' to ensure adhered liquid is returned to the flask. Flat head allows stopper to be stood on end, thus preventing contamination of the ground surface.
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Hollow-blown to reduce weight