Tisk…
Jars with knobbed lid, DURAN®
Dodavatel: DWK Life Sciences
242050109 242050306 242050503 242051002 242052101 242053209
216-3360EA 1920 CZK
216-3360 216-3361 216-3362 216-3363 216-3364 216-3365
Jars with knobbed lid, DURAN®
Nádoby Vzorkové sklenice
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Lid has polished rim for easy handling.