Tisk…
Tanks, cylindrical, with cover, Nalgene®
Dodavatel: Thermo Fisher Scientific
11200-0005 11200-0007 11200-0010 11200-0015 11200-0030 11200-0055 11200-0100
216-1900EA 9690 CZK
216-1900 216-1901 216-1902 216-1903 216-1904 216-1905 216-1906
Tanks, cylindrical, with cover, Nalgene®
Zásobníky Zásobníky válcové
These cylindrical polypropylene tanks with cover can resist stress cracking and increase tolerance to many organic chemicals.
  • An economic alternative to stainless-steel tanks
Informace pro objednávání: Factory-installed spigots and tank liners are sold separately.
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí NAR Certificate of Retirement