Tisk…
Crystallising dishes, DURAN®
Dodavatel: DWK Life Sciences
213112401 213113209 213113809 213114102 213114402 213114908 213115407 213115904 213116309 213132403 213133408 213133802 213134104 213134404 213134901 213135409 213135906 213136302 213113406 213133202
216-1810EA 448 CZK
216-1810 216-1811 216-1813 216-1814 216-1815 216-1816 216-1817 216-1818 216-1819 216-1820 216-1862 216-1863 216-1864 216-1865 216-1866 216-1867 216-1868 216-1869 216-1812 216-1821
Crystallising dishes, DURAN®
Podnosy/Misky/Kelímky Misky krystalizační
DURAN®, borosilicate glass 3.3, clear, with or without spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For a wide range of applications. From 100 ml capacity upwards has white square labelling area.
*without labelling field
DIN 12338 (with spout)
Certifikace: DIN 12337 (without spout)
DIN 12338 (with spout)