Tisk…
Quickstart universal containers, PP, 30 ml, STERILIN®
Dodavatel: Sterilin
128A/P 128BBAC/P 128B/P 128C/P
216-1395EA 0 CZK
216-1395 216-1397 216-1396 216-1398
Quickstart universal containers, PP, 30 ml, STERILIN®
Nádobky na vzorky Nádoby pro obecné použití
Polypropylene, ultra clear, with QickStart cap.
  • Aseptically manufactured
  • Suitable for centrifugation at 9500 ×g
  • Multi-seal cap 'QuickStart' for improved handling and unrivaled leak-free performance
  • Lot number printed on each container to aid full traceability
  • Labeled for patient details and available either undosed, dosed with boric acid preservative (0,4 g) or a loose spoon
Secure excellent temperature and chemical resistance with these 95kPa compliant and CE marked leak-resistant universal containers.
Certifikace: Leak-tested in accordance withMore Product Information