Tisk…
Evaporating dishes, DURAN®
Dodavatel: DWK Life Sciences
213013202 213013408 213013802 213014104 213014404 213014901 213015409 213015906 213016302
216-1270EA 292 CZK
216-1270 216-1271 216-1272 216-1273 216-1274 216-1275 216-1276 216-1277 216-1278
Evaporating dishes, DURAN®
Podnosy/Misky/Kelímky Misky odpařovací
DURAN®, borosilicate glass 3.3, clear, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For a wide range of applications. With flat bottom and (from volume 170 ml upwards) matt labelling area.
*without labelling field
Certifikace: DIN 12336