Tisk…
Buckets, round, Nalgene®
Dodavatel: Thermo Fisher Scientific
7002-0025
216-0093EA 3560 CZK
216-0093
Buckets, round, Nalgene®
Kbelíky
HDPE, white with spout.
  • Tough and durable
  • Graduated in pins and litres
  • Fitted with polethylene coated wire handle
Ideal for transfer and transport of laboratory solutions and powders.