Tisk…
Láhve, úzké hrdlo, kulaté, HDPE, se šroubovacím uzávěrem, Nalgene®
Dodavatel: Thermo Fisher Scientific
2002-0001 2002-0002 2002-0004 2002-0006 2002-0008 2002-0016 2002-0032 2002-9016 2002-9025 2002-9050 2002-9125
215-7504EA 455 CZK
215-7504 215-7505 215-7506 215-7507 215-7508 215-7510 215-7511 215-7509 215-7502 215-7503 215-7501
Láhve, úzké hrdlo, kulaté, HDPE, se šroubovacím uzávěrem, Nalgene®
Láhve Láhve plastové
HDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant up to +100 °C
  • Leakproof
Ideal for storing and transporting liquids. Suitable for a wide range of applications.
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact,Product Transparency and Disclosure
Certifikát ochrany životního prostředí ISO14001 (Certificate Registration No. EMS 752603), ACT Label
NAR Certificate of Retirement
NAR Certificate of Retirement NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603, ACT Label NAR Certificate of Retirement, ACT Label