Tisk…
Measuring cylinders, tall form, borosilicate glas, Class B, DURAN®
Dodavatel: DWK Life Sciences
213965405 213966307 213963601 213960707 213960801 213961403 213961703 213962408 213964409
215-3646EA 1870 CZK
215-3646 215-3647 215-3644 215-3639 215-3640 215-3641 215-3642 215-3643 215-3645
Measuring cylinders, tall form, borosilicate glas, Class B, DURAN®
Válce Válce odměrné
DURAN® borosilicate glass 3.3, clear, Class B.
  • Calibrate to contain ('In')
  • Marks and inscription in white
  • With spout
  • With large hexagonal base
Base ist equipped with three knobs to increase the stability
Certifikace: DIN ISO 4788