Tisk…
Pipette baskets, Nalgene®
Dodavatel: Thermo Fisher Scientific
DS5241-0020 DS5241-0040 DS5241-0050
215-0032EA 8030 CZK
215-0032 612-1802 215-0034
Pipette baskets, Nalgene®
Koše a košíky Košíky na pipety
Made of HDPE. Chemical resistant pipette baskets protect glass pipettes from breakage.
  • Rugged, resilient baskets cushion pipettes and won't corrode
  • Chemically compatible with most acids, bases, alcohols and detergents
A perforated screen in the bottom allows cleaning solutions to fill the chamber from the bottom and is recessed 1,9 cm for complete drainage. Use these high denisty polyethylene baskets inside Nalgene™ pipette jars to soak pipettes in cleaning or disinfection solutions, then drain and transfer into the Nalgene™ pipette washer forMore Product Information