Tisk…
PE stoppers, DURAN®
Dodavatel: DWK Life Sciences
292050707 292050201 292050304 292050604 292050801 292050904 292051103 292051206 292050407
214-0399EA 439 CZK
214-0399 214-0395 214-0396 214-0398 214-0400 214-0401 214-0402 214-0403 214-0397
PE stoppers, DURAN®
Zátky Zátky PE/LDPE/HDPE
These PE stoppers have an ergonomic grip that makes the opening and closing of the flasks easier and more secure. Coloured inserts in the different sizes of stoppers allow the easy identification of the corresponding ground (NS/TS) neck size.
Certifikace: DIN 12254