Tisk…
Measuring beakers with handle, coloured
Dodavatel: VITLAB
481943 481944 481945 4811111 481942 4821111 482944 482942 482943 482945
213-3750EA 1460 CZK
213-3750 213-3751 213-3752 213-3753 213-3749 VITL4821111 VITL482944 VITL482942 VITL482943 VITL482945
Measuring beakers with handle, coloured
Kádinky
PP, transparent.
  • Stackable
  • Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions
  • Available in different colours
Ideal for food stuffs. With readable printed scale.