Tisk…
Measuring beakers with handle
Dodavatel: VITLAB
442081 443081 444081 446081 447081 445081 440081 441081
213-3725EA 166 CZK
213-3725 213-3730 213-3735 213-3705 213-3710 213-3740 213-3715 213-3720
Measuring beakers with handle
Kádinky
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use and a wide variety of other applications. With embossed scale, blue graduations.
Food-safe products acc. to EC Directive No. 10/2011.